Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Disposable Cleanroom Socks
GREENER_CHOICE

Catalog No. 128960036 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
11 to 15
6 to 10
Color:
White, Blue Stitching
White, Red Stitching
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
12-896-0036 11 to 15 White, Red Stitching
12-896-0035 6 to 10 White, Blue Stitching
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-896-0036
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Disposable cleanroom socks are specialized for critical environments and are a durable, cost-effective option for cleanroom and extended wear.

  • Fisherbrand Disposable Cleanroom Socks are a cost-effective and cleanroom compatible option for cleanroom and extended wear.
  • Laundered and packaged in ISO Class 5 Cleanrooms, with extremely low particulate contamination
  • The structure of the socks allows high wear-resistance and durability while maintaining good elasticity.
  • Abrasion-resistant, soft, and comfortable to wear.
  • The product should be stored in a ventilated, dry, mildew-proof and insect-proof place.
  • Avoid prolonged product exposure to sunlight.
  • Do not rub against rough objects directly.
  • Launder using a neutral or slightly alkaline (pH <9) synthetic detergent on a gentle cycle to prevent mechanical and chemical damage
  • If the socks are torn or the yarn begins to falls off or tear in multiple places, replace them.

Specifications

Certifications/Compliance ISO Class 5 (Class 100)
Color White, Red Stitching
For Use With (Application) Cleanroom and other critical environments
Gender Unisex
Height (Metric) 50 cm
Length (Metric) 26 cm
Material Polyester
Size 11 to 15
Weight (Metric) 25 g/pair
Disposable Single-use
Sterility Non-sterile
Packaging Quantity 500 Pair/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.