Learn More
Radiometer America Radiometer ABL90 FLEX PLUS Capillary Clot Catcher (250 units)
SDP
Catalog No. NC1210611
Encompass_Preferred
ABL90 FLEX PLUS Clot Catcher (250 units)
Disposable Clot Catcher for ABL 90 FLEX blood gas analyzer, 250 units per package
ABL90 FLEX PLUS Blood Gas Analyzer
The ABL90 FLEX PLUS analyzer gives you reliable results. With the ABL90 FLEX analyzer you get lab quality test results. In just 35 seconds, our cassette-based ABL90 FLEX analyzer can deliver 17 parameters from a sample as small as 65 μL. ABL90 measures blood gas, electrolytes, metabolites and co-oximetry. The ABL90 analyzer can measure a sample from a syringe, capillary tube or test tube.
Catalog No. NC1210611 Supplier Radiometer America Supplier No. 906026
ABL90 FLEX PLUS Clot Catcher (250 units)
Disposable Clot Catcher for ABL 90 FLEX blood gas analyzer, 250 units per package
ABL90 FLEX PLUS Blood Gas Analyzer
The ABL90 FLEX PLUS analyzer gives you reliable results. With the ABL90 FLEX analyzer you get lab quality test results. In just 35 seconds, our cassette-based ABL90 FLEX analyzer can deliver 17 parameters from a sample as small as 65 μL. ABL90 measures blood gas, electrolytes, metabolites and co-oximetry. The ABL90 analyzer can measure a sample from a syringe, capillary tube or test tube.