Learn More
TA Instruments Disposable Concentric Cylinder Cups, Aluminum, Package of 20, Nominal Diameter 32 mm, for Use with P/N 545050.901 Kit
Catalog No. 5000580879 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Disposable Concentric Cylinder Cups are aluminum cups packaged in sets of 20. They are for use with the disposable concentric cylinder kit (P/N 545050.901) and have a nominal diameter of 32 mm.
Catalog No. 50-005-80879 Supplier TA Instruments Supplier No. 545075.901
Waters TA Instruments Disposable Concentric Cylinder Cups are aluminum cups packaged in sets of 20. They are for use with the disposable concentric cylinder kit (P/N 545050.901) and have a nominal diameter of 32 mm.