Learn More
TA Instruments Disposable Concentric Cylinder Kit for Peltier and Electrically Heated Jackets, Includes Cup Holder, for Cups P/N 545075.901, Temperature up to 150 Degrees Celsius
Catalog No. 5000580660 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Disposable Concentric Cylinder Kit For Peltier And Electrically Heated Jackets includes a cup holder necessary for using disposable cups (P/N 545075.901) with Peltier or Electrically Heated Concentric Cylinder Jackets up to 150 degrees Celsius. Disposable cups are not included with the kit.
Catalog No. 50-005-80660 Supplier TA Instruments Supplier No. 545050.901
Waters TA Instruments Disposable Concentric Cylinder Kit For Peltier And Electrically Heated Jackets includes a cup holder necessary for using disposable cups (P/N 545075.901) with Peltier or Electrically Heated Concentric Cylinder Jackets up to 150 degrees Celsius. Disposable cups are not included with the kit.