Learn More
Lakeland Industries Disposable Coveralls
Supplier:  Lakeland Industries TG4124X
Encompass_Preferred
MicroMax adds a durable polyester scrim for the strength and protection you need from dirt, dust, grease, grime and light chemical splash. Open wrists and ankle for easy donning and doffing.
Catalog No. 50-284-0998