Learn More
Lakeland Industries Disposable Coveralls
Catalog No. 502841054
Encompass_Preferred
SafeGard SMMS Features: ? Global offering ? great for multinational companies ? Global standardized testing and certificatio (CE Certified Type 5/6) ? 4 sizing options simplify ordering and reduces inventory requirements? (SM/MD, LG/XL, 2X/3X, 4X/5X) ? Serged seams ? Anti-Stat (EN1149-1) ? Comfortable, highly breathable fabric ? Excellent particle filtration efficiency
Catalog No. 50-284-1054 Supplier Lakeland Industries Supplier No. SSG417B2X3X
SafeGard SMMS Features: ? Global offering ? great for multinational companies ? Global standardized testing and certificatio (CE Certified Type 5/6) ? 4 sizing options simplify ordering and reduces inventory requirements? (SM/MD, LG/XL, 2X/3X, 4X/5X) ? Serged seams ? Anti-Stat (EN1149-1) ? Comfortable, highly breathable fabric ? Excellent particle filtration efficiency