Learn More
Lakeland Industries Disposable Coveralls
Supplier:  Lakeland Industries SSG428BSMMD
Encompass_Preferred
SafeGard SMMS Features:\n?Global offering ? great for multinational companies Global standardized testing and certificatio (CE Certified Type 5/6) 4 sizing options simplify ordering and reduces inventory requirements (SM/MD, LG/XL, 2X/3X, 4X/5X) Serged seams Anti-Stat (EN1149-1) Comfortable, highly breathable fabric Excellent particle filtration efficiency
Catalog No. 50-284-1055