Learn More
Lakeland Industries Disposable Coveralls
Supplier:  Lakeland Industries TG4174X
Encompass_Preferred
MicroMax adds a durable polyester scrim for the strength and protection you need from dirt, dust, grease, grime and light chemical splash. Elastic wrists and ankle for optimal protection.
Catalog No. 50-284-1064