Learn More
Lakeland Industries Disposable Coveralls
Supplier:  Lakeland Industries TG4145X
Encompass_Preferred
MicroMax adds a durable polyester scrim for the strength and protection you need from dirt, dust, grease, grime and light chemical splash. Attached hood and boots for maximum coverage and protection.
Catalog No. 50-284-0754