Learn More
Lakeland Industries Disposable Coveralls
Supplier:  Lakeland Industries TG428MD
Encompass_Preferred
MicroMax adds a durable polyester scrim for the strength and protection you need from dirt, dust, grease, grime and light chemical splash. Attached hood with elastic wrists and ankle for optimal protection.
Catalog No. 50-284-1069