Learn More
Lakeland Industries Disposable Coveralls
Catalog No. 502841127
Encompass_Preferred
Lightweight, breathable protection from Lakeland's MicroMax NS products. This coverall features an attached hood and boots with elastic wrists. CTL414 is ideal for protection against dirt, grease, grime, and light chemical splash.
Catalog No. 50-284-1127 Supplier Lakeland Industries Supplier No. CTL414V5X
Lightweight, breathable protection from Lakeland's MicroMax NS products. This coverall features an attached hood and boots with elastic wrists. CTL414 is ideal for protection against dirt, grease, grime, and light chemical splash.