Learn More
Lakeland Industries Disposable Coveralls
Supplier:  Lakeland Industries CTL414VLG
Encompass_Preferred
Lightweight, breathable protection from Lakeland's MicroMax NS products. This coverall features an attached hood and boots with elastic wrists. CTL414 is ideal for protection against dirt, grease, grime, and light chemical splash.
Catalog No. 50-284-1128