Learn More
Lakeland Industries Disposable Coveralls
Catalog No. 502840994
Encompass_Preferred
Lakeland's SafeGard SMMS products provide economical protection from dry particulate and light liquid splash protection while streamlining a product offering across our global markets, making this garment a perfect fit for international companies. The SSG414B configuration features an attached hood and boots and elastic wrists. Dual-sizing allows for more streamlined ordering and inventory management.
Catalog No. 50-284-0994 Supplier Lakeland Industries Supplier No. SSG414B2X3X
Lakeland's SafeGard SMMS products provide economical protection from dry particulate and light liquid splash protection while streamlining a product offering across our global markets, making this garment a perfect fit for international companies. The SSG414B configuration features an attached hood and boots and elastic wrists. Dual-sizing allows for more streamlined ordering and inventory management.