Learn More
Lakeland Industries Disposable Coveralls
Supplier:  Lakeland Industries SSG414BLGXL
Encompass_Preferred
Lakeland's SafeGard SMMS products provide economical protection from dry particulate and light liquid splash protection while streamlining a product offering across our global markets, making this garment a perfect fit for international companies. The SSG414B configuration features an attached hood and boots and elastic wrists. Dual-sizing allows for more streamlined ordering and inventory management.
Catalog No. 50-284-1093