Learn More
Sigma Aldrich Fine Chemicals Biosciences Disposable culture tubes p
Catalog No. 502900185
Encompass
Ideal for use in bacteriology RIA coagulation and other routine laboratory procedures. Polypropylene tubes can be centrifuged at 3000AxAg and sterilized at 120C. Polystyrene tubes can be centrifuged at 1400AxAg but can not be autoclaved.
Catalog No. 50-290-0185 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3767951PAK
Ideal for use in bacteriology RIA coagulation and other routine laboratory procedures. Polypropylene tubes can be centrifuged at 3000AxAg and sterilized at 120C. Polystyrene tubes can be centrifuged at 1400AxAg but can not be autoclaved.