Learn More
Thomas Scientific DISPOSABLE CUVETTES, ACRYLIC
SDP
Catalog No. NC1939107
Encompass_Preferred
Disposable Cuvettes, Acrylic (UV grade, useable to 275nm), Volume: 1.5ml, 2 Clear
Catalog No. NC1939107 Supplier Thomas Scientific Supplier No. 1158A08
Disposable Cuvettes, Acrylic (UV grade, useable to 275nm), Volume: 1.5ml, 2 Clear