Learn More
IMPLEN INC Implen DiluCell™ 10 disposable cuvettes, pack of 96, for standard 1 cm cuvette spectrophotometers
Catalog No. NC3404476
Encompass_Preferred
Plastics, Cuvettes DiluCell™ 10 DiluCell™ 10 disposable cuvettes, pack of 96, Previously it was necessary to use standard cuvettes with a volume of at least 1 ml and additionally to dilute the sample manually. DiluCell™ from Implen allows for lower sample volume requirements and provides automatic virtual sample dilution; Due to the unique design of DiluCell™ cuvettes, the light-path is reduced from the standard 10 mm to 1.0 mm (DC 10); According to the Lambert Beer Law this shortened pathlength results in an automatic sample dilution by factor of 10; Automatic dilution with DiluCell™ saves time and excludes dilution errors and cross contaminations; Low sample volume necessary; Suitable for proteins and microbial cell cultures; Measurement possible starting at 340 nm; Automatic dilution of factor 10; No risk of contamination; Significantly delayed cell sedimentation compared to standard cuvettes
Catalog No. NC3404476 Supplier IMPLEN INC Supplier No. DC10
Plastics, Cuvettes DiluCell™ 10 DiluCell™ 10 disposable cuvettes, pack of 96, Previously it was necessary to use standard cuvettes with a volume of at least 1 ml and additionally to dilute the sample manually. DiluCell™ from Implen allows for lower sample volume requirements and provides automatic virtual sample dilution; Due to the unique design of DiluCell™ cuvettes, the light-path is reduced from the standard 10 mm to 1.0 mm (DC 10); According to the Lambert Beer Law this shortened pathlength results in an automatic sample dilution by factor of 10; Automatic dilution with DiluCell™ saves time and excludes dilution errors and cross contaminations; Low sample volume necessary; Suitable for proteins and microbial cell cultures; Measurement possible starting at 340 nm; Automatic dilution of factor 10; No risk of contamination; Significantly delayed cell sedimentation compared to standard cuvettes