Learn More
World Precision Instrument Disposable Dissection Boards-WP-630-1
SDP
Catalog No. 502538393
Encompass
The DispoCut™ Disposable Dissection Board is strong but inexpensive enough to throw away. It is a clean, safer and more efficient way of handling infectious tissue specimens. Ideal for pathology labs, medical schools, classroom dissections, forensic labs, medical research labs, pharmaceutical labs, surgery and more. When sterilizing, DispoCut™ may be safely wiped down with alcohol or autoclaved. Reversible.
Catalog No. 50-253-8393 Supplier World Precision Instrument Supplier No. WP6301
The DispoCut™ Disposable Dissection Board is strong but inexpensive enough to throw away. It is a clean, safer and more efficient way of handling infectious tissue specimens. Ideal for pathology labs, medical schools, classroom dissections, forensic labs, medical research labs, pharmaceutical labs, surgery and more. When sterilizing, DispoCut™ may be safely wiped down with alcohol or autoclaved. Reversible.