Learn More
Pyramex™ Disposable Ear Plugs
Catalog No. 19555144 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Corded:
Corded
Uncorded
Packaging Type:
Box
Dispenser bin
Retail Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded Packaging Type
19-555-144 Uncorded Dispenser bin
19-555-143 Uncorded Retail Pk.
19-155-294 Uncorded Box
19-155-295 Corded Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-555-144 Supplier Pyramex™ Supplier No. PYDP1000BN
Add to Cart
Add to Cart
Soft disposable foam ear plugs gently expand and self adjust to all size ear canals
  • Contoured fit disposable polyurethane ear plugs
  • Gently expands and self adjusts to all size ear canals
Specifications
ANSI S3.1974 Yes
Size Universal
For Use With (Application) Assembly, Construction, Manufacturing, Transportation, Utilities, and Other Various Industries, and Other Various Industries
High Visibility Yes
Packaging Type Dispenser bin
Pairs per Package 1000 Pairs/Bin
Plug Material Foam
Plug Color Orange
Noise Reduction Rating (NRR) 32 dB
Corded Uncorded
Plug Shape Bullet
Product Type Disposable Earplugs
Disposable Single-use
Metal Detectable No
Dielectric Yes