Learn More
MARIAN MEDICAL INC Disposable ENFit Syringe, 100 ml, 30 Per Box
Catalog No. NC3958227
Encompass_Preferred
Syringe. Disposable Syringe. Marian Medical Inc. ME100. Disposable syringe for single patient use. Manufactured to ISO 80369 3 connection standards. Sterile individually packaged syringe. Clear barrel with easy to read graduations. For medication administration in clinical and home care settings. 30 per box.
Catalog No. NC3958227 Supplier MARIAN MEDICAL INC Supplier No. ME100
Syringe. Disposable Syringe. Marian Medical Inc. ME100. Disposable syringe for single patient use. Manufactured to ISO 80369 3 connection standards. Sterile individually packaged syringe. Clear barrel with easy to read graduations. For medication administration in clinical and home care settings. 30 per box.