Learn More
Fisherbrand™ Disposable Face Mask
Catalog No. 12888001C Shop All Fisherbrand Products
Change view
Click to view available options
Face Mask with Earloops, 3-ply., CE Certification, FDA Certification and GB/T 32610-2016, Blue, Disposable, Latex Free, Polypropylene, 500/Cs., Non-Sterile
Catalog No.
12888001C Face Mask with Earloops, 3-ply., CE Certification, FDA Certification and GB/T 32610-2016, Blue, Disposable, Latex Free, Polypropylene, 500/Cs., Non-Sterile
1 options
Catalog No. 12888001C
3-ply Face masks. Latex and fiberglass free.
  • 3-Ply material
  • Latex and Fiberglass fiber free
  • Malleable noseband ensures adjustable seal
  • Non-woven pleated
  • CE Mark
Specifications
Product Type General Purpose Face Mask
Color Blue
Material Polypropylene
Sterility Non-sterile
Certifications/Compliance CE Mark
Disposable Single-use
Latex Free Yes
Model 12-888-001C
Packaging Quantity 500/Cs.