Learn More
Medicus Health Disposable Face Shield With Drape, Extra Long, 30 Pk
SDP
Supplier:  Medicus Health 2547M4
Encompass
∙ Disposable Face Shields with Extra Long Protective Drape (30 Pack) are Anti-Fog, Comfortable, Latex-Free, Provide Excellent Personal Protection against Face Splash and come with a Fluid Barrier Fabric that Extends 13" ∙Crystal-clear, anti-fog plastic with patented flat viewing area in front of the eye is anti-glare and designed to minimize visual distortion∙Manufactured under the highest quality, CE Marked and OSHA level protection against face splash∙ 30 Face Shields per pack∙Extra Long Fluid Barrier can be tucked inside of a sterile gown, providing additional protection against blood splash coming up under the shield∙Also designed to be worn freely outside a gown protecting the upper neck, chin and mouth from splash coming under the shield∙Light pressure on the integral hook and loop fasteners on the bottom of the barrier allows for option closure so the fabric is tight under the chin∙Meets OSHA’s bloodborne pathogen regulations
Catalog No. NC1155331