Learn More
Braintree Scientific Disposable Feeding Needle w/Bite Protector,Sterile, 18G-1.5""
SDP
Catalog No. 501954341
Encompass
Flexible Disposable Plastic (PTFE) Needles with Bite Protectors Flexible Plastic (PTFE) tube is covered at the hub end with 304 stainless steel tubing this protects tubing from damage due to biting during use. Plastic (PTFE) tube is softer than other tubing types, very flexible, and translucent for easy confirmation of reagent delivery. Plastic luer hubs and soft silicone tips, non-toxic and non-pyrogenic. Packaged 5 needles per sterile peel pouch/20 pouches per box. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4341 Supplier Braintree Scientific Supplier No. BG9928
 Restricted Item
Add to Cart
Add to Cart
Flexible Disposable Plastic (PTFE) Needles with Bite Protectors Flexible Plastic (PTFE) tube is covered at the hub end with 304 stainless steel tubing this protects tubing from damage due to biting during use. Plastic (PTFE) tube is softer than other tubing types, very flexible, and translucent for easy confirmation of reagent delivery. Plastic luer hubs and soft silicone tips, non-toxic and non-pyrogenic. Packaged 5 needles per sterile peel pouch/20 pouches per box. **Sold for research purposes only and not for human use**