Learn More
Lakeland Industries Disposable FR Coverall
Supplier:  Lakeland Industries 511305X
Encompass_Preferred
Lakeland Pyrolon CRFR coveralls are suitable when moderate chemical exposure and flash fire are a concern, generally used in paint manufacturing, environmental clean-up, or oil and gas. 51130 is a secondary FR coverall configured with a hood, elastic wrists, and ankles. Coming in a case of 6, with sizes SM-5X available, Pyrolon CRFR is designed to protect primary FR against flammable contaminants and dry particulates.
Catalog No. 50-284-0787