Learn More
Lakeland Industries Disposable FR Coverall
Supplier:  Lakeland Industries 521324X
Encompass_Preferred
Lakeland Pyrolon CBFR coveralls are suitable for most advanced chemical protection when flash fire is present, meeting ANSI/ISEA 203.  52132 is a secondary FR coverall configured with a respirator fit hood, elastic wrists, and ankles. Coming in a case of 6, with sizes MD-5X available, Pyrolon CBFR is designed for the highest chemical hold out to protect primary FR clothing, and your people.
Catalog No. 50-284-0908