Learn More
Lakeland Industries Disposable FR Coverall
Supplier:  Lakeland Industries 517304X
Encompass_Preferred
Lakeland Pyrolon CRFR aprons are suitable when moderate chemical exposure and flash fire are a concern, generally used in paint manufacturing, environmental clean-up, or oil and gas.  51730 is a secondary FR apron configured with sewn tied, elastic wrists, and long sleeves. Coming in a case of 12, with sizes SM-5X available, Pyrolon CRFR is designed to protect primary FR against flammable contaminants and dry particulates.
Catalog No. 50-284-1162