Learn More
Miles Scientific CALIBRATED MICROPIPETTES - 10UL (PKG 100)
SDP
Catalog No. NC0583446
Encompass_Preferred
Calibrated end to end disposable glass micropipettes provide economical, precise measurement of TLC samples with better than 1% accuracy. The tubes are quickly filled by capillary action or with the spotting bulb. One spotting bulb is supplied with each vial of 100 micropipettes. 
Catalog No. NC0583446 Supplier Miles Scientific Supplier No. A2010
Calibrated end to end disposable glass micropipettes provide economical, precise measurement of TLC samples with better than 1% accuracy. The tubes are quickly filled by capillary action or with the spotting bulb. One spotting bulb is supplied with each vial of 100 micropipettes. 