Learn More
Sklar Instruments Large Sklar Disposable Graves Vaginal Specula, Sklar . Non-Sterile. Latex-Free. CS/100.
SDP
Catalog No. NC0790457 Shop All Sklar Instruments Products
Encompass_Preferred
Large SklarSpec Disposable Graves Vaginal Specula, Sklar. Non-Sterile. Latex-Free. SklarSpec plastic specula are clear, disposable and comfortable. Plastic material allows 360 degree visibility and reduces thermal shock. Provides protection for the doctor, the nurse and the patient from cross-contamination. Clean-up is eliminated by single-use only. One hand operation with wide range adjustment. - CS100.
Catalog No. NC0790457 Supplier Sklar Instruments Supplier No. 960877
Large SklarSpec Disposable Graves Vaginal Specula, Sklar. Non-Sterile. Latex-Free. SklarSpec plastic specula are clear, disposable and comfortable. Plastic material allows 360 degree visibility and reduces thermal shock. Provides protection for the doctor, the nurse and the patient from cross-contamination. Clean-up is eliminated by single-use only. One hand operation with wide range adjustment. - CS100.