Description
- Ideal for anaerobic microbiology applications where deep agar inoculation is required
- Use in situations where flaming is not practical, such as under hoods and in anaerobic chamber
- High-impact polystyrene loops are smooth and flexible to facilitate smooth, uniform streaking without damaging the gel surface
- Rigid enough to penetrate the media for subsurface inoculation
- Disposable loops do not need flaming, thus help to eliminate the risk of infection due to aerosol formation of pathogenic substances
- Can be used under hoods without danger
- Color-coded for ease of size identification
Specifications
Specifications
| Disposable | Single-use |
| Volume | 1 μL |
| Material | Polystyrene |
| Sterility | Sterile |
| Color | Blue |
| Quantity | Cs of 1000 (40 Pks. of 25) |
| Autoclavable | Not Autoclavable |