Learn More
GVS Disposable Inoculating Loop
Catalog No. 09317341 Shop All GVS Products
Change view
Click to view available options
Volume:
1 μL
10 μL
10 μL, 1 μL
Color:
Blue
Dark Blue
Natural
Neutral
White
Yellow
Material:
ABS
AS
High-impact Polystyrene
Polypropylene
Packaging Type:
1/Peel Pack
10/Zip-Lock Pack
20/Zip-Lock Pack
5/Zip-Lock Pack
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume Color Material Packaging Type
09-317-341 10 μL, 1 μL Yellow AS 5/Zip-Lock Pack
09-316-825 10 μL Blue Polypropylene 20/Zip-Lock Pack
09-316-797 10 μL Yellow ABS 10/Zip-Lock Pack
09-317-040 10 μL Blue Polypropylene 10/Zip-Lock Pack
09-316-758 10 μL, 1 μL Yellow AS 1/Peel Pack
09-316-738 10 μL, 1 μL Yellow ABS 20/Zip-Lock Pack
09-316-739 10 μL Blue Polypropylene 1/Peel Pack
09-316-997 1 μL Neutral Polypropylene 20/Zip-Lock Pack
09-317-169 10 μL, 1 μL Yellow ABS 5/Zip-Lock Pack
09-317-170 10 μL, 1 μL Yellow ABS 1/Peel Pack
09-317-168 1 μL Neutral Polypropylene 10/Zip-Lock Pack
09-317-147 10 μL, 1 μL Yellow AS 20/Zip-Lock Pack
09-317-172 10 μL, 1 μL Yellow AS 10/Zip-Lock Pack
09-317-181 1 μL Neutral Polypropylene 1/Peel Pack
09-317-179 10 μL Blue Polypropylene 5/Zip-Lock Pack
09-317-177 1 μL Natural Polypropylene 5/Zip-Lock Pack
09-316-308 10 μL Dark Blue High-impact Polystyrene 10/Zip-Lock Pack
09-316-256 10 μL Yellow High-impact Polystyrene 10/Zip-Lock Pack
09-316-140 10 μL Dark Blue High-impact Polystyrene 5/Zip-Lock Pack
09-316-141 10 μL Dark Blue High-impact Polystyrene 1/Peel Pack
09-316-391 1 μL White High-impact Polystyrene 1/Peel Pack
09-316-500 1 μL White High-impact Polystyrene 5/Zip-Lock Pack
09-316-643 1 μL White High-impact Polystyrene 10/Zip-Lock Pack
09-316-642 1 μL White High-impact Polystyrene 20/Zip-Lock Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. 09-317-341 Supplier GVS Supplier No. CELCUCG2121X0051S17
Add to Cart
Add to Cart
GVS Disposable Inoculating Loops and Needles available in 1 and 10 μL with different packing for choices.
  • Smooth Surface – Ensures even and consistent streaking
  • Color-Coded Sizes – Allows quick and easy identification
  • Ergonomic Polygonal Shaft – Provides a secure grip for easy operation and precise directional control
  • Loop Options – Available in both rigid and flexible types upon request
  • Secure Packaging – Supplied in individual peel packs (easy peel-open) and zip-lock packs (tamperproof and re-sealable) to minimize the risk of contamination
Specifications
Material AS
Sterility Sterile
Volume 10 μL, 1 μL
Size Standard
Color Yellow
Product Type Inoculating Loop
Packaging Type 5/Zip-Lock Pack