Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Disposable Inoculating Needles

Catalog No. 07000806 Shop All Greiner Bio-One Products
Change view
Click to view available options
Quantity:
1 Ea.
2000
3000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-000-806 3000
07-000-805 2000
07-000-283 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-806 Supplier Greiner Bio-One Supplier No. 731185
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in picking single colonies. Greiner Bio-One Disposable Inoculating Needles are flexible for collection and inoculation.

  • Sterilised by irradiation
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Length (English) 7.87 in.
Material Polystyrene
Length (Metric) 200 mm
Sterility Sterile
Size Standard
Product Type Inoculating Needle
Quantity 3000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.