Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Essentials Disposable Inoculation Needles

Catalog No. p-7076781
Change view
Click to view available options
Quantity:
40
100
250
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-391-557 40
03-391-558 100
03-391-563 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-391-557 Supplier Cole-Parmer™ Essentials Supplier No. 0623109
Only null left
Add to Cart
Add to Cart

Save time and eliminate cross-contamination

Smooth and flexible needles are constructed of high-impact polystyrene. Straight needles are ideal for picking single colonies. Color coding identifies inoculation volume easily. Packed sterile in tamperproof resealable bags. Each case is supplied with a quality certificate confirming sterility and accuracy.

Specifications

Disposable Single-use
Length (English) 9 in.
Length (Metric) 22.7 cm
Sterility Sterile
Material Polystyrene
Color White
Product Type Inoculating Needle
Quantity 40
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.