Learn More
Electron Microscopy Sciences Disposable Knife
SDP
Catalog No. 501928487
Encompass_Preferred
This poly-handled disposable knife's blade is a size 40, non-sterile and made of carbon steel.
Catalog No. 50-192-8487 Supplier Electron Microscopy Sciences Supplier No. 7205487
This poly-handled disposable knife's blade is a size 40, non-sterile and made of carbon steel.