Learn More
Lakeland Industries Disposable Lab Coats
Catalog No. 502840625
Encompass_Preferred
MicroMax NS provides the protection you need in the laboratory from dirt, grease, grime, light chemical splash and blood. Long sleeve design has open wrists and a snap front.
Catalog No. 50-284-0625 Supplier Lakeland Industries Supplier No. CTL1405X
MicroMax NS provides the protection you need in the laboratory from dirt, grease, grime, light chemical splash and blood. Long sleeve design has open wrists and a snap front.