Learn More
Lakeland Industries Disposable Lab Coats
Supplier:  Lakeland Industries CTL1402X
Encompass_Preferred
MicroMax NS provides the protection you need in the laboratory from dirt, grease, grime, light chemical splash and blood. Long sleeve design has open wrists and a snap front.
Catalog No. 50-284-1207