Learn More
BURKLE INC DispoLadle SteriPlast, HDPE, sterile, 39.37 in. (1000 mm)
Catalog No. 503081321
Encompass_Preferred
DispoLadle is a disposable sampling scoop specially designed for accurate liquid sampling. The scaled scoop features embossed measurements in 10 ml (0.34 oz.) increments, up to 200 ml (6.76 oz.), ensuring precise volume control.
Manufactured in a cleanroom environment, individually packaged, and gamma-sterilized, DispoLadle meets the highest hygiene standards. Its ergonomic handle and dual spouts allow easy handling for both right- and left-handed users, making liquid sampling simple and efficient.
Catalog No. 50-308-1321 Supplier BURKLE INC Supplier No. 53936651
DispoLadle is a disposable sampling scoop specially designed for accurate liquid sampling. The scaled scoop features embossed measurements in 10 ml (0.34 oz.) increments, up to 200 ml (6.76 oz.), ensuring precise volume control.
Manufactured in a cleanroom environment, individually packaged, and gamma-sterilized, DispoLadle meets the highest hygiene standards. Its ergonomic handle and dual spouts allow easy handling for both right- and left-handed users, making liquid sampling simple and efficient.