Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ Disposable Manual 55mL Reagent Reservoirs
GSA_VA

Catalog No. 12111087 Shop All OHAUS Products
Change view
Click to view available options
Packaging:
1 reservoir per bag
Bulk
No. per Pack:
100 reservoirs/Pk.
20 bags/Pk.
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging No. per Pack Sterility
12-111-087 Bulk 100 reservoirs/Pk. Non-sterile
12-111-086 1 reservoir per bag 20 bags/Pk. Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-111-087 Supplier BIOTIX™ Supplier No. 63300122
Get Extra Pipette Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed with angled sides and V-shaped bottom.

  • Made from polystyrene
  • With graduation marks

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Translucent
Capacity (Metric) 55 mL
Material Polystyrene
Sterility Non-sterile
Type Reservoir
For Use With (Application) Filling Multi-channel Pipettors
No. per Pack 100 reservoirs/Pk.
Packaging Bulk
Quantity 100/Cs.
Autoclavable Autoclavable
Channels 12
Disposable Single-use
Bottom Shape Flat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.