Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TerraCycle™ Disposable Masks Zero Waste Box™
SDPSCI_ED

Catalog No. p-7203480
Change view
Click to view available options
Capacity (Metric):
39.66 L
79.31 L
136.42 L
Dimensions (L x W x H):
11 x 11 x 20 in.
11 x 11 x 40 in.
15 x 15 x 37 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Dimensions (L x W x H)
S88854115 39.66 L 11 x 11 x 20 in.
S88854116 79.31 L 11 x 11 x 40 in.
S88854117 136.42 L 15 x 15 x 37 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options

These boxes are not suitable for medical waste. This includes any red bag waste (i.e. material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.) The zero waste box system does not accept household items that may be considered hazardous waste. Do not ship dripping packages.

Not available in Hawaii, Alaska, and Puerto Rico.

Catalog No. S88854115 Supplier TerraCycle™ Supplier No. CS83S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These boxes are not suitable for medical waste. This includes any red bag waste (i.e. material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.) The zero waste box system does not accept household items that may be considered hazardous waste. Do not ship dripping packages.

Not available in Hawaii, Alaska, and Puerto Rico.

Recycle disposable masks. Each box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

  • Select a Zero Waste Box for a specific waste stream and place your order.
  • Collect the accepted items to fill the system.
  • Send it back to TerraCycle with the prepaid return label on your system.
  • Order your next Zero Waste Box system to continue to recycle.
  • Includes: One Zero Waste Box system, One prepaid return label affixed to your box, Outbound shipping to your location, Processing and recycling of the materials, Customer support from TerraCycle's team of experts
  • Types of masks that can be included: disposable face masks, including N95, KN95, 3-ply nonwoven, and dust masks; cannot include paper masks or reusable fabric masks.

Order Info

This product is not available in Hawaii, Alaska, or Puerto Rico.

Specifications

Capacity (Metric) 39.66 L
Closure Type Fold-down flap
Color Brown, Green, White
Dimensions (L x W x H) 11 x 11 x 20 in.
Height (Metric) 50.8 cm
Length (Metric) 27.94 cm
Shape Rectangle
Width (Metric) 27.94 cm
Material Paperboard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.