Learn More
Electron Microscopy Sciences Disposable Micro Tissue, 0.75 ml
SDP
Catalog No. 501900371
Encompass_Preferred
The disposable micro tissue grinder allows for homogenized tissue micro samples without cross contamination. This unit is especially useful for radioactive samples. No washing is needed; just disposed the set after use. Mortar Size: 10 x 44 mm. Pestle Size: 7 mm sphere. Made from Autocalvable Wheaton 33 low extractable borosilicate glass.
Catalog No. 50-190-0371 Supplier Electron Microscopy Sciences Supplier No. 6480307
Add to Cart
Add to Cart
The disposable micro tissue grinder allows for homogenized tissue micro samples without cross contamination. This unit is especially useful for radioactive samples. No washing is needed; just disposed the set after use. Mortar Size: 10 x 44 mm. Pestle Size: 7 mm sphere. Made from Autocalvable Wheaton 33 low extractable borosilicate glass.