Learn More
Cole-Parmer Disposable HDPE Milling Jars, 0.125 gal (0.5 L); 12/Pk
Supplier:  Cole-Parmer 0417200
Encompass
Disposable milling jars eliminate cross-contamination problems. Jars handle loads as small as 75 grams—perfect for research applications. Baffled jars prevent grinding media from slipping and provide highly efficient tumbling action. Made from high-density polyethylene (HDPE), these jars withstand strong chemicals. Jar sleeves sold separately.
Catalog No. NC2017114