Learn More
Grainger Adam Equipment Disposable Moisture Analyzer Pans, Aluminum, 10 mm, 250/Pk.
Catalog No. 502794729
Encompass_Preferred
These disposable aluminum pans are designed for use with moisture analyzers to facilitate accurate moisture analysis testing. The pans provide a stable, contained surface for samples, helping to keep materials securely in place during analysis.
  • Pack quantity: 250 pans
  • Material: Aluminum
  • Depth: 10 mm
  • Finish: Smooth
  • Compatibility: Suitable for use with moisture analyzers
Catalog No. 50-279-4729 Supplier Grainger Supplier No. 19YN80
These disposable aluminum pans are designed for use with moisture analyzers to facilitate accurate moisture analysis testing. The pans provide a stable, contained surface for samples, helping to keep materials securely in place during analysis.
  • Pack quantity: 250 pans
  • Material: Aluminum
  • Depth: 10 mm
  • Finish: Smooth
  • Compatibility: Suitable for use with moisture analyzers