Learn More
MSC IronClad Disposable Nitrile Glove, Orange, 8 Mil, XL, 100/Pk.
Catalog No. 5000686587
Encompass
These disposable nitrile gloves serve as a general-use barrier against liquids, grease, and dirt. Designed for professional use, they offer an alternative solution for individuals allergic to natural rubber latex.
  • Provides protection from unwanted and dangerous substances
  • Features a beaded cuff to ensure easy donning and prevent roll down
  • Textured surface enhances wet and dry gripping
  • Offers strength and puncture resistance
  • Compliant with FDA regulations
  • Hypoallergenic construction
  • Suitable for automotive, safety, sanitary, medical, tattoo, laboratory, and manufacturing industries
Catalog No. 50-006-86587 Supplier MSC Supplier No. 68950534
These disposable nitrile gloves serve as a general-use barrier against liquids, grease, and dirt. Designed for professional use, they offer an alternative solution for individuals allergic to natural rubber latex.
  • Provides protection from unwanted and dangerous substances
  • Features a beaded cuff to ensure easy donning and prevent roll down
  • Textured surface enhances wet and dry gripping
  • Offers strength and puncture resistance
  • Compliant with FDA regulations
  • Hypoallergenic construction
  • Suitable for automotive, safety, sanitary, medical, tattoo, laboratory, and manufacturing industries