Learn More
Sigma Aldrich Fine Chemicals Biosciences Disposable PD 10 Desalting
Catalog No. 502904193
Encompass
Disposable PD-10 Desalting Columns offer an inexpensive and convenient alternative to lengthy and troublesome dialysis procedures. In minutes biological samples are desalted and small molecular weight solutes are removed or exchanged with excellent recoveries of macromolecules.The instructions for Disposable PD-10 Desalting Columns have been updated with two new application protocols gravity or centrifugation to increase the flexibility of the product. The gravity protocol allows a simple cleanup of samples with one or several columns in parallel without the need for a purification system. The spin protocol enables runs in parallel in a standard centrifuge with minimal dilution of the eluted sample.four adapters are included in each product pack to enable easy centrifugation. Together with LabMate PD-10 Buffer Reservoir wash and equilibration buffer can be applied in one step with the gravity flow protocol. Empty PD-10 columns are recommended for use with bulk media for affordable
Catalog No. 50-290-4193 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17085101
Disposable PD-10 Desalting Columns offer an inexpensive and convenient alternative to lengthy and troublesome dialysis procedures. In minutes biological samples are desalted and small molecular weight solutes are removed or exchanged with excellent recoveries of macromolecules.The instructions for Disposable PD-10 Desalting Columns have been updated with two new application protocols gravity or centrifugation to increase the flexibility of the product. The gravity protocol allows a simple cleanup of samples with one or several columns in parallel without the need for a purification system. The spin protocol enables runs in parallel in a standard centrifuge with minimal dilution of the eluted sample.four adapters are included in each product pack to enable easy centrifugation. Together with LabMate PD-10 Buffer Reservoir wash and equilibration buffer can be applied in one step with the gravity flow protocol. Empty PD-10 columns are recommended for use with bulk media for affordable