Learn More
Med Vet International Disposable Pediatric Anesthesia Circuit With 1 Liter Bag, J0248B
Catalog No. 5000582221
Encompass_Preferred
For use with smaller patients, this lessens the amount of dead space. Consists of two 30" breathing tubes (5/8"), inhaler "Y" piece and mask elbow. Clear color.
Catalog No. 50-005-82221 Supplier Med Vet International Supplier No. JOR248B
For use with smaller patients, this lessens the amount of dead space. Consists of two 30" breathing tubes (5/8"), inhaler "Y" piece and mask elbow. Clear color.