Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Disposable PES Filter Units
GREENER_CHOICE

Catalog No. p-7113064 Shop All Fisherbrand Products
Change view
Click to view available options
Pore Size:
0.20 μm
0.45 μm
Volume (Metric):
150 mL
250 mL
500 mL
1000 mL
Diameter (Metric) Membrane:
50 mm
75 mm
90 mm
For Use With (Application):
Fluid clarification and particle removal
Sterile Filtration of media and serum
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Volume (Metric) Diameter (Metric) Membrane For Use With (Application)
FB12566500 0.20 μm 150 mL 50 mm Sterile Filtration of media and serum
FB12566501 0.45 μm 150 mL 50 mm Fluid clarification and particle removal
FB12566502 0.20 μm 250 mL 50 mm Sterile Filtration of media and serum
FB12566503 0.45 μm 250 mL 50 mm Fluid clarification and particle removal
FB12566504 0.20 μm 500 mL 75 mm Sterile Filtration of media and serum
FB12566505 0.45 μm 500 mL 75 mm Fluid clarification and particle removal
FB12566506 0.20 μm 1000 mL 90 mm Sterile Filtration of media and serum
FB12566507 0.45 μm 1000 mL 90 mm Fluid clarification and particle removal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. FB12566500
Only null left
Add to Cart
Edge
Add to Cart

Includes

Individually packaged sterile screw top style bottle cap

Increase cell culture sample protection with Fisherbrand sterile disposable vacuum filter units with PES membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.

  • PES membrane is low protein binding to maintain protein balance
  • PES membrane has low extractables to maintain media purity
  • Upper chamber of filter units is clearly marked with pore size, membrane type, catalog number, lot number and expiration date
  • Non-pyrogenic and non-cytotoxic
  • Gamma radiation-sterilized and individually bagged for a 5-year sterile shelf life
  • 0.2μm units for sterile filtration
  • 0.45μm units for fluid clarification and particle removal

Certifications

Non-pyrogenic per USP Standard Lot-to-Lotnon-cytotoxic Lot-to-Lot sterility

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity Low binding
Certifications/Compliance Non-pyrogenic per USP Standard Lot-to-Lot
Disposable Single-use
Material PES
Material (Membrane) PES
Pore Size 0.20 μm
Sterility Sterile
For Use With (Application) Sterile Filtration of media and serum
Type Filter Unit
Volume (Metric) 150 mL
Diameter (Metric) Membrane 50 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.