Learn More
KIMBLE™ Disposable PESTLE™ Replacement Microtube, natural PP
SDP
Catalog No. 01672740 Shop All DWK Life Sciences Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
01-672-740 Autoclavable
1 options
Catalog No. 01-672-740 Supplier KIMBLE™ Supplier No. 7495600500
Add to Cart
Add to Cart
Replacement microtubes constructed of polypropylene.
  • Manufactured from polypropylene
  • Autoclavable
  • Disposable
  • Pestles specially designed to match microtubes
  • Can be driven by cordless motor 749540-0000
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Homogenization of Tissue Samples
Autoclavable Autoclavable
Capacity (Metric) 0.5 mL