Learn More
Research Products International Corp Disposable Petri Dishes, 100 x 15mm, Sterile, 25 per Sleeve, 500 per Case
SDP
Catalog No. 50998465
Encompass
Sterile Petri Dishes allow easy one hand operation with flared lid skirt and squared corners. Dishes feature ventillation ribs that allow air circulation; reduced condensation during incubation; and consistently flat surface for even media distribution. Manufactured from medical grade virgin polystyrene. Available in various styles, rims,compartments and designs.
Ideal for automation or general use
Optically clear
Medical Grade
Catalog No. 50-998-465 Supplier Research Products International Corp Supplier No. 160268
Sterile Petri Dishes allow easy one hand operation with flared lid skirt and squared corners. Dishes feature ventillation ribs that allow air circulation; reduced condensation during incubation; and consistently flat surface for even media distribution. Manufactured from medical grade virgin polystyrene. Available in various styles, rims,compartments and designs.
Ideal for automation or general use
Optically clear
Medical Grade