Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Disposable Plastic Base Molds
SDP

Catalog No. 21010262
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-010-262 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-010-262 Supplier StatLab Supplier No. BM1515
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Single use, clear plastic base molds.

  • Eliminates use of cleaning solvents and release compounds
  • 500/Cs.
  • Approximately 17 x 17 x 5 mm

Specifications

Product Type Base Mold
Material Plastic
Color Clear
Length (Metric) 17 mm
Width (Metric) 17 mm
Dimensions (L x W x H) 17 x 17 x 5 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.