Learn More
Sigma Aldrich Fine Chemicals Biosciences Disposable Plastic BioSafe
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z740303150EA
Encompass
Eliminate Cross ContaminationThe first pipette with total contamination controlThe Argos BioSafe pipette is a standard serological pipette with a unique barrier filter. If over-aspiration occurs with BioSafe pipettes liquid is locked and unable to pass through to filter. Similar in theory to a barrier pipette tip BioSafe pipettes provide sample integrity while promoting user safety. It also protects electronic pipettors from contamination and cross contaminating samples by over aspiration.Ideal in applications involving any potentially hazardous or dangerous biological fluids infectious diseases or when cross over contamination is an issue.Unique filter plug locks liquid in pipette during over-aspirationAvailable in 5 10 25 mL (individually wrapped)Eliminate aerosol or liquid contaminationTotal contamination protection for pipettor and samplesUse in place of filtered pipette tips
Catalog No. 50-290-0850