Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Disposable Plastic Cuvets
SCI_ED

Catalog No. S29159 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
4.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S29159 4.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S29159

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

  • Disposable 10 mm pathlength cuvets designed for visible and UV/Vis assays
  • Select polystyrene cuvets for assays in visible spectral range (340 to 750 nm)
  • For measurements in the visible and near-UV range (300-750 nm) choose cuvets made from polymethyl methacrylate (acrylic)
  • 4.5 sample capacity
  • Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
  • Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5 mL
  • Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity

 

Order Info

Packaged 100 cuvets per stackable tray; five trays to a case; Outside dimensions: 12L x 12W x 45mmH

Specifications

Capacity (Metric) 4.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.